Tri-Sil TP (TMSI:Pyridine) Reagent
Product Image
Tri-Sil TP (TMSI:Pyridine) Reagent
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-49231
also known as PIERCE #49231
25 mL
TPCTS-4923010 x 1 mL ampules
TS-49230
also known as PIERCE #49230
10 x 1 mL ampules
TPCTS-4923125 mL
Catalog number TS-49231
also known as PIERCE #49231
Price (MYR)
-
Quantity:
25 mL
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent. Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.
Additonal Product Information
Does not react with amines
May be used in the presence of water as long as there is enough reagent present to react with both the water and the sample.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithGC
Standard Concentration65 - 80, 20 - 25%
HazardsFlammable
Item Description25 mL
Storage2°C to 8°C
Quantity25 mL
Standard ComponentsPyridine, 1-(Trimethylsilyl)-1 H-imidazole
Unit SizeEach