Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent
Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent
Actual product may vary
Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-49001
also known as PIERCE #49001
50 mL
TPCTS-4899910 x 1 mL ampules
TS-48999
also known as PIERCE #48999
10 x 1 mL ampules
TPCTS-4900150 mL
Catalog number TS-49001
also known as PIERCE #49001
Price (MYR)
-
Quantity:
50 mL
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent, Composed of HMDS, TMCS and high-purity pyridine, it rapidly produces TMS derivatives of polar compounds for GC analysis and biochemical synthesis. This versatile reagent is ideally suited for GC determinations of a range of compounds including sugars, alcohols, phenols, steroids, sterols, organic acids, and some amines.
  • Derivatizes carbohydrates, phenols, steroids, sterols, organic acids, alcohols and some amines
  • Useful for rapid production of TMS derivatives of polar compounds for gas chromatographic determination and biochemical synthesis
  • Versatility, speed and ease of use of Tri-Suil HTP Reagent has made it the most widely used silylation formulation
For Research Use Only. Not for use in diagnostic procedures.
General References:
  1. Ng, L., et al. (1993). J. Chromatogr. 637, 104-108.
Specifications
For Use WithGC
Standard Concentration65 - 80, 10 - 20, 5 - 7%
HazardsCorrosive, Flammable
Item Description50 mL
Storage2°C to 8°C
Quantity50 mL
Standard ComponentsPyridine, Hexamethyldisilazane, Chlorotrimethylsilane
Unit SizeEach