MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent
MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent
Actual product may vary
MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent
Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStandard Components
TS-48920
also known as PIERCE #48920
MTBSTFA
TPCTS-48920MTBSTFA
Catalog number TS-48920
also known as PIERCE #48920
Price (MYR)
-
Standard Components:
MTBSTFA
Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.
Additional Product Information:
  •  Derivatives have a high molecular concentration at M-57.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithGC
Standard Concentration98 to 100%
HazardsIrritant
Storage2°C to 8°C
Quantity5 mL ampules
Standard ComponentsMTBSTFA
Unit SizeEach