Water-Soluble Siliconizing Fluid
Water-Soluble Siliconizing Fluid
Actual product may vary
Water-Soluble Siliconizing Fluid
To siliconize glass surfaces use Thermo Scientific™ Water-Soluble Siliconizing Fluid.
Have Questions?
Catalog NumberQuantity
TS-42799
also known as PIERCE #42799
120 mL
Catalog number TS-42799
also known as PIERCE #42799
HSN 90189044
Price (MYR)
0.00
Each
Add to cart
Quantity:
120 mL
Price (MYR)
0.00
Each
Add to cart
To siliconize glass surfaces use Thermo Scientific™ Water-Soluble Siliconizing Fluid. This easy-to-use silane monomer solution is supplied as a 20% solid solution in a mixture of diacetone alcohol and tertiary butyl alcohol. The primary silane component is an octadecyltrialkoxysilane, which is hydrolyzed to a silanol when mixed with water. It provides greater resistance to base hydrolysis than other surface treatments.
  • Easy-to-use silane monomer solution that is supplied as a 20% solid solution in a mixture of diacetone alcohol and tertiary butyl alcohol
  • Greater resistance to base hydrolysis than other surface treatments
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithSiliconizing Glass Surfaces
HazardsFlammable, Irritant
Standard Components4-Hydroxy-4-Methylpentan-2-One, 2-Methyl-2-Propanol, Silane, Trimethoxyoctadecyl
Standard Concentration25 - 45, 25 - 45, 10 - 20%
Item Description120 mL
Quantity120 mL
Storage15°C to 35°C
Product TypeWater-Soluble Siliconizing Fluid
Unit SizeEach