BSTFA Silylation Reagent
BSTFA Silylation Reagent
Actual product may vary
BSTFA Silylation Reagent
Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-38830
also known as PIERCE #38830
10 x 1 mL ampules
TPCTS-3882825 g
TS-38828
also known as PIERCE #38828
25 g
TPCTS-38829100 g
TS-38829
also known as PIERCE #38829
100 g
TPCTS-3883010 x 1 mL ampules
Catalog number TS-38830
also known as PIERCE #38830
Price (MYR)
-
Quantity:
10 x 1 mL ampules
Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
BSTFA is a powerful thrimethylsilyl donor, with donor strength that is comparable to its unfluorinated analog BSA [N,O-Bis(trimethylsilyl)acetamide]. BSTFA reacts to replace labile hydrogens on a wide range of polar compounds with a -Si(CH3)3 group. This physical characteristic is particularly useful in the gas chromatography of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
  • Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA
  • Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA
For Research Use Only. Not for use in diagnostic procedures.
General References:
  1. Her, G.R., et al. (1985). Anal. Biochem. 151, 292-298.
  2. Shanchun, J., et al. (1994). Org. Geochem. 21, 415-422.
Specifications
For Use WithGC
Standard Concentration98 to 100%
Item Description10 x 1 mL ampules
Storage2°C to 8°C
Quantity10 x 1 mL ampules
Standard ComponentsTrimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
Unit SizeEach