Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Actual product may vary
Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Use these triangular stir bars for faster reaction times and smooth mixing in vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Description
TPRTS-160003, 5, and 10 mL Reacti-Vials6 Pack
TPRTS-160100.3 and 1mL Reacti-Vials6 Pack
Catalog number TPRTS-16000
Price (MYR)
1,370.00
PK
Add to cart
For Use With (Equipment):
3, 5, and 10 mL Reacti-Vials
Description:
6 Pack
Price (MYR)
1,370.00
PK
Add to cart
Use these triangular stir bars for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminum Blocks.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description6 Pack
For Use With (Equipment)3, 5, and 10 mL Reacti-Vials
TypeMagnetic Stirrer
Unit SizePK