Tri-Sil TBT (TMSI:BSA:TMCS) Reagent
Product Image
Tri-Sil TBT (TMSI:BSA:TMCS) Reagent
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent.
Have Questions?
Catalog NumberQuantity
TPCTS-4901610 x 1 mL ampules
Catalog number TPCTS-49016
Price (MYR)
4,046.00
Pack
Add to cart
Quantity:
10 x 1 mL ampules
Price (MYR)
4,046.00
Pack
Add to cart
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent. Composed of TMSI/BSA/TMCS (3:3:2), this catalyzed silylation reagent formulation typically reacts with hydroxyls in a short period of time at 60–80 °C to form TMS ethers. Highly hindered hydroxyls may require several hours.
  • Converts all classes of hydroxyl groups to TMS ethers
  • Under usual conditions, the reaction is complete in a short period of time at 60 to 80°C, although very hindered hydroxyls may require several hours
For Research Use Only. Not for use in diagnostic procedures.
General References:
  1. Seidel, V., et al. (1993). Chromatographia 37, 191-201.
Specifications
For Use WithGC
Standard Concentration25 - 45, 25 - 45, 20 - 25%
HazardsToxic
StorageRoom Temperature
Quantity10 x 1 mL ampules
Standard ComponentsTrimethylsilyl N-trimethylsilylacetamidate, 1-(Trimethylsilyl)-1 H-imidazole, Chlorotrimethylsilane
Unit SizePack