RVCR Series 12 in. Class 100/ISO 5 Nitrile Cleanroom Gloves
RVCR Series 12 in. Class 100/ISO 5 Nitrile Cleanroom Gloves
Actual product may vary
RVCR Series 12 in. Class 100/ISO 5 Nitrile Cleanroom Gloves
Rival CR's 12 in. soft modulus nitrile gloves deliver the fit and feel of latex for the ultimate stretch and comfort. Suitable for Class 100 compatible cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
TNGRVCR1504X-Large
TNGRVCR1501Small
TNGRVCR1502Medium
TNGRVCR1503Large
Catalog number TNGRVCR1504
Price (MYR)
1,247.00
CS
Add to cart
Size:
X-Large
Price (MYR)
1,247.00
CS
Add to cart
  • 100% nitrile, does not contain natural rubber or silicone
  • Suitable for Class 100 compatible cleanroom, pharmaceutical, biotech and medical device manufacturing
  • 12 in. length, soft modulus, accelerator free
  • Meets the most stringent requirements for extractables and particles
  • Microtextured fingertips provide and secure grip on all surfaces
Specifications
ChlorinatedYes
Cleanroom ClassClass 100 (ISO 5)
Accelerator FreeYes
AQL1.5
ColorWhite
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)5 mil
Finger Thickness (Metric)0.15 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4 mil
Palm Thickness (Metric)0.12 mm
PowderedPowder Free
Product TypeControlled Environment Gloves
SizeX-Large
SterilityNon-sterile
Thickness (English)4 mil
Thickness (Metric)0.10 mm
Manufacturing/QMS Audit StandardsISO9001:2000 and ISO 14001:2004
ModelRVCR1504
Packaging TypeCleanroom Bagged
SeriesRVCR
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS