Owl™ Transfer Membranes
Owl™ Transfer Membranes
Actual product may vary
Owl™ Transfer Membranes
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging Type
TM151-5Sheets (10 x 10cm)
TM151-1Roll
TM151-4Sheets (15 x 15cm)
Catalog number TM151-5
Price (MYR)
2,386.00
EA
Add to cart
Packaging Type:
Sheets (10 x 10cm)
Price (MYR)
2,386.00
EA
Add to cart
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
  • Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™.
  • This membrane provides the highest signal-to-noise ratio, which allows the use of mild blocking procedures.
  • Immobilon-P has high protein retention, is compatible with all standard visualization methods and can be used for protein sequencing.
  • Immobilon is available in minigel and large-format gel sizes.
Specifications
Length (Metric)10 cm
Width (Metric)10 cm
Packaging TypeSheets (10 x 10cm)
Description10 x 10cm; Sheets
For Use With (Application)Western blotting
FormatSheet
MaterialImmobilon-P, PVDF
Dimensions (LxW)10 cm x 10 cm
Product LineOwl
Unit SizeEA