Lindberg/Blue M™ Mini-Mite™ Tube Furnaces
Lindberg/Blue M™ Mini-Mite™ Tube Furnaces
Actual product may vary
Lindberg/Blue M™ Mini-Mite™ Tube Furnaces
Quickly heat up and/or cool down samples with this convenient and flexible portable single-tube furnace, insulated with Thermo Scientific™ Moldatherm™.
Have Questions?
Catalog NumberController TypeElectrical Requirements
TF55030C-1
also known as AVL
Digital, Single Segment208/240 V 50/60 Hz
Catalog number TF55030C-1
also known as AVL
Price (MYR)
17,918.00
EA
Add to cart
Controller Type:
Digital, Single Segment
Electrical Requirements:
208/240 V 50/60 Hz
Price (MYR)
17,918.00
EA
Add to cart
Quickly heat up and/or cool down samples with this convenient and flexible portable Thermo Scientific™ Lindberg/Blue M™ Mini-Mite™ Tube Furnaces, insulated with Moldatherm. Microprocessor-based self-tuning PID control provides optimal thermal processes without overshooting.
Key Features:
  • Single segment, single setpoint, one ramp to setpoint
  • Adjustable high-limit overtemperature protection
  • Simultaneous LED display of temperature and setpoint in °C or °F
  • Split-hinge design simplifies loading and unloading
  • Safety switch disconnects power when furnace is opened
  • Type K long-life thermocouple
Recommended for:
  • Pyrolisis
  • Thermal Expansion
  • Calibration
  • Sintering
  • Viscoisty Testing
Includes:
  • 9'(3m) power cord
Warranty:
  • 12 months
Specifications
Controller TypeDigital, Single Segment
Heated Zone305 mm
Temperature (Metric)100°C to 1100°C
TypeSingle Segment
DisplayLED
Power Consumption800 w
Dimensions (L x W x H) Exterior11 x 16 x 15 in. (28 x 41 x 38 cm)
Length (English)11 in.
Length (Metric)28 cm
Width (English)16 in.
Width (Metric)41 cm
Height (English)15 in.
Height (Metric)38 cm
Diameter (English) Process Tube1 in.
Diameter (Metric) Process Tube2.54 cm
Shipping Weight (English)35 lb.
Shipping Weight (Metric)16 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V