Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Actual product may vary
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Ensure consistently reliable and accurate fluid transfers. This durable high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerThickness (English)
TBP8600-03103/4 in.1/8 in.
FIS14-179-1111/8 in.1/16 in.
FIS14-179-1243/4 in.1/8 in.
Catalog number TBP8600-0310
Price (MYR)
3,806.00
EA
Add to cart
Diameter (English) Inner:
3/4 in.
Thickness (English):
1/8 in.
Price (MYR)
3,806.00
EA
Add to cart
Ensure consistently reliable and accurate fluid transfers with Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing for Peristaltic Pumpsis.This flexible, durable high-purity translucent tubing is designed for a variety of pump and transfer applications, including pharmaceutical, laboratory and bioprocess manufacturing.
  • Tasteless, odorless, non-cytotoxic and non-hemolytic
  • Contains no phthalate plasticizers or peroxides that can leach out into transported fluid
  • Low binding suface to minimize absorption of proteins and food products
  • Autoclavable; also can be sterilized by gamma radiation, EtO and chemical procedures
  • Durometer hardness: Shore A, 50
Ordering Information:  Double-bagged in case of 50 ft. (15.2m)
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceMeets USP<87> Cytotoxicity (Agar Diffusion and MEM Elution methods); USP<88> Class VI; USP<661>Physicochemical Tests; ISO10993-3 Bacterial Mutagenicity – AmesAssay; ISO10993-4 Hemolysis Direct Contact, EP 3.1.9 Silicone Elastomer f
AutoclavableAutoclavable
Diameter (English) Inner3/4 in.
Diameter (Metric) Inner19 mm
MaterialPlatinum-cured Silicone
Diameter (English) Outer1 in.
Diameter (Metric) Outer25.4 mm
Overall Length (English)50 ft.
Thickness (English)1/8 in.
Thickness (Metric)3.2 mm
ColorTranslucent
Hardness50 (Shore A)
Flexibility4 x OD bend radius
Maximum Pressure9 psig at 73°F (23°C)
Product TypeFlexible Tubing
Unit SizeEA