Smart-Tracker™ Wireless Datalogging Module
Smart-Tracker™ Wireless Datalogging Module
Actual product may vary
Smart-Tracker™ Wireless Datalogging Module
Ensuring that the cold chain has been preserved, whether your products are in storage or in transit, just makes sense. Now make it simple.
Have Questions?
Catalog NumberDescriptionFor Use With (Application)Temperature Range (Metric)Calibration
ST100-102Smart-Tracker wireless datalogging module with internal temperature probeFreezers, refrigerators, transportation, cold storage-40°C to +85°C-30°C
Catalog number ST100-102
Price (MYR)
2,176.00
EA
Add to cart
Description:
Smart-Tracker wireless datalogging module with internal temperature probe
For Use With (Application):
Freezers, refrigerators, transportation, cold storage
Temperature Range (Metric):
-40°C to +85°C
Calibration:
-30°C
Price (MYR)
2,176.00
EA
Add to cart
Ensuring that the cold chain has been preserved, whether your products are in storage or in transit, just makes sense. Thermo Scientific™ Smart-Tracker™ makes temperature monitoring in the cold chain easy. All you need is a smart phone to wirelessly check stored temperature readings. There is no dedicated equipment, no docking stations and no complex computer installations.
Download the Smart-Tracker mobile app: iPhoneAndroid
  • Track temperature wirelessly and control modules using your smart phone with the Smart-Tracker application
  • Monitor storage facilities, freezers, refrigerators, heating equipment and packages in transit†
  • Share information via the Cloud anywhere in the world
  • Bluetooth wireless connection for use in many types of container and enclosures
  • Internal sensor: -20° to +60°C or -40° to +85°C
  • External PT100 sensor: -200° to +200°C
  • Warranty varies by country, please contact us for details
† Subject to network speed and availability
Specifications
TypeWireless Datalogging Module with Internal Probe
Data Memory4000 Readings
Calibration-30°C
Software TypeiPhone 4S, iPhone 5, iPhone 6, iPad 3⁄4, Android
Battery TypeLithium
DescriptionSmart-Tracker wireless datalogging module with internal temperature probe
IncludesModule with internal sensor, lithium battery, two colored sensor rings and two side magnetic mount
Certifications/ComplianceBluetooth Certification: FCC, CE
ModelST100-102
Sensor TypesInternal Sensor, External PT100 Sensor
For Use With (Application)Freezers, refrigerators, transportation, cold storage
Temperature Range (Metric)-40°C to +85°C
Dimensions (L x W x H)0.24 x 0.63 x 1.02 in. (6 x 16 x 26 mm) [Shipping Dimensions]
Shipping Weight (English)0.44 lb.
Unit SizeEA