Isotemp™ Advanced Hotplate Stirrer, Ambient to 450°C, Ceramic
Isotemp™ Advanced Hotplate Stirrer, Ambient to 450°C, Ceramic
Actual product may vary
Isotemp™ Advanced Hotplate Stirrer, Ambient to 450°C, Ceramic
Isotemp™ Advanced Hotplate Stirrer, Ambient to 450°C, Ceramic
Isotemp™ Advanced Hotplate Stirrer, Ambient to 450°C, Ceramic
Isotemp™ Advanced Hotplate Stirrer, Ambient to 450°C, Ceramic
Stirring Range: 50 to 1500 rpm, Shape: Square, Size: 7.25 x 7.25 in., Plug Type: EU and UK
Have Questions?
Catalog NumberPlug Type
SP88857295UK/EU/CN
Catalog number SP88857295
Price (MYR)
5,090.00
EA
Add to cart
Plug Type:
UK/EU/CN
Price (MYR)
5,090.00
EA
Add to cart
Reliable performance, dependable safety and precise control. Available with optimized controls and settings for applications that demand advanced precision.
Intuitive design:
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Ability to save temperature setting
  • Includes stainless-steel PT100 temperature probe, stir bar, supporting rod and clamp kit
  • Optional splash guard protection shield
Advanced technology:
  • StirTrac features innovative engineering for slow-speed stirring, consistent speed control, and stronger magnetic coupling
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Temperature adjustable in 1°C increments except when using temperature probe then ability to control in 0.1 degree increments below 100°C
  • EU and UK plug
Specifications
Top Plate MaterialCeramic
Voltage230 V
DescriptionHotplate Stirrer
Electrical Requirements230 V 50/60 Hz
Length (English)8.2 in.
Size7.25 x 7.25 in.
Height (Metric)9.7 cm
Temperature (Metric)Ambient to 450°C
Height (English)3.8 in.
Frequency50/60 Hz
Width (Metric)33 cm
Length (Metric)20.8 cm
Width (English)13 in.
TypeStirring Hotplate
ShapeSquare
Plug TypeUK/EU/CN
Max. Container Size4 L
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (LxWxH)7.25 in. x 7.25 in.
Certifications/ComplianceRoHS, cCSAus (100 to 120 V Model), CE (230 V Model)
Unit SizeEA