Check Order Status
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
Actual product may vary
Fisherbrand
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
Have Questions?
Change view
Catalog Number
Quantity
SP120-500
500 mL
FCMSP120-500
500 mL
FCMSP120-1
1 L
3 Options
Catalog number
SP120-500
Price (MYR)
721.00
EA
Add to cart
Quantity:
500 mL
Price (MYR)
721.00
EA
Add to cart
Specifications
Color
Yellow
pH Range
1.1
Format
Liquid
Packaging
Glass Bottle
Quantity
500 mL
Type
Platinum Cobalt Color Standard Solution