Eccentric Luer-Slip Tip Three-Part Syringe
Eccentric Luer-Slip Tip Three-Part Syringe
Actual product may vary
Eccentric Luer-Slip Tip Three-Part Syringe
Terumo™ Eccentric Luer-Slip Tip Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
Have Questions?
Catalog NumberVolume (Metric)Product TypeQuantity
SMCSS30LE130 mLLuer-Lock Tip 3-Part Syringe50/Pk.
Catalog number SMCSS30LE1
Price (MYR)
215.00
BX
Add to cart
Volume (Metric):
30 mL
Product Type:
Luer-Lock Tip 3-Part Syringe
Quantity:
50/Pk.
Price (MYR)
215.00
BX
Add to cart
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Eccentric Luer-Tip Three-Part Syringe feature a unique double beveled design for greater patient comfort.
  • Silicone coated barrel for a smooth plunger action
  • Clear barrel to allow quick recognition of air bubbles
  • Easy to read markings
  • Latex free
  • Sterile and packed in individually wrapped tamper peel packs
  • Syringes are available with luer lock, luer slip, catheter and eccentric tips
  • Available in a wide range of sizes
Specifications
MaterialPolypropylene
DescriptionWithout Needle
Product TypeLuer-Lock Tip 3-Part Syringe
DisposableSingle-use
Quantity50/Pk.
Volume (Metric)30 mL
Connection TypeLuer Lock
GraduatedYes
SterilitySterile
Unit SizeBX