Check Order Status
Needle Hypodermic
Needle Hypodermic
Needle Hypodermic
Have Questions?
Catalog Number
Needle Gauge
Length (Metric) Needle
Quantity
SMCAN2038R1
20 G
38 mm
100 Pk.
Catalog number
SMCAN2038R1
Price (MYR)
20.00
BX
Add to cart
Needle Gauge:
20 G
Length (Metric) Needle:
38 mm
Quantity:
100 Pk.
Price (MYR)
20.00
BX
Add to cart
Needle Hypodermic
Agani Thin Wall
Large bore hypodermic intervention is especially useful in catastrophic blood loss or shock.
A hypodermic needle is used for rapid delivery of liquids, or when the injected substance cannot be ingested, either beca
Specifications
Material
Stainless Steel
Color
Yellow
Needle Gauge
20 G
Length (English) Needle
1.5 in.
Product Type
Hypodermic Needle
Disposable
Single-use
Quantity
100 Pk.
Length (Metric) Needle
38 mm
Needle Point Style
Sharp
For Use With (Equipment)
Syringe
Sterility
Non-sterile
Unit Size
BX