Agani™ 18 G Thin Wall Hypodermic Needle
Agani™ 18 G Thin Wall Hypodermic Needle
Actual product may vary
Agani™ 18 G Thin Wall Hypodermic Needle
Thin wall hypodermic needle with pink needle hub, suitable for all applications.
Have Questions?
Catalog NumberNeedle Gauge
SMCAN1838R118 G
Catalog number SMCAN1838R1
Price (MYR)
20.00
BX
Add to cart
Needle Gauge:
18 G
Price (MYR)
20.00
BX
Add to cart
Thin wall hypodermic needle with pink needle hub, suitable for all applications.
Terumo™ Hypodermic Needles are latex-free and precision manufactured for greater patient comfort.
  • Individually packaged in a sterile, color-coded blister pack for easy gauge identification
  • Reassurance provided by visualization of flashback through semi-transparent color-coded needle hub
  • Thin wall needles allow you to choose a smaller gauge size with increased flow rates over regular walled needles
  • Suitable for all applications
Specifications
ColorPink
Needle Gauge18 G
Latex FreeYes
Product TypeHypodermic Needle
Quantity100 Pk.
Length (Metric) Needle38 mm
Unit SizeBX