MaxQ™ Digital Shakers PROMO
Product Image
MaxQ™ Digital Shakers PROMO
Provides reliable results required for bacteria and yeast cultures.
Have Questions?
Catalog NumberModelHoldsProduct Type
SHKE4450GB-1CEMaxQ 4450250 mL flask clampsIncubated Shaker
Catalog number SHKE4450GB-1CE
Price (MYR)
35,260.00
EA
Add to cart
Model:
MaxQ 4450
Holds:
250 mL flask clamps
Product Type:
Incubated Shaker
Price (MYR)
35,260.00
EA
Add to cart
  • Designed to provide reliable results required for bacteria and yeast cultures
  • Durable triple eccentric drive
  • Ten year parts warranty
  • Maximise capacity of 250mL and 2L flasks in compact footprint
  • Digital display with simultaneous viewing of temperature, speed and time with audible and visual alarms
Specifications
Holds250 mL flask clamps
Product TypeIncubated Shaker
Controller TypeDigital
ModelMaxQ 4450
Includes1 x 330 x 279mm universal platform, 9 x 250mL flask clamps
Motor TypeTriple eccentric drive
Platform StyleUniversal platform
SizeSmall
Unit SizeEA