MaxQ™ 4000 Gassing Manifold
MaxQ™ 4000 Gassing Manifold
Actual product may vary
MaxQ™ 4000 Gassing Manifold
Enable direct gassing into flasks for anaerobic cultures with the Thermo Scientific™ MaxQ™ 4000 Gassing Manifold.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
SHK4000-8GMGassing manifold with 8 outletsMaxQ 4000
Catalog number SHK4000-8GM
Price (MYR)
6,709.00
EA
Add to cart
Description:
Gassing manifold with 8 outlets
For Use With (Equipment):
MaxQ 4000
Price (MYR)
6,709.00
EA
Add to cart
Enable direct gassing into flasks for anaerobic cultures with the Thermo Scientific™ MaxQ™ 4000 Gassing Manifold. With exceptional innovation, reliability and accuracy, MaxQ™ 4000 shakers, platforms and accessories are the optimal choice for a busy lab.
Specifications
Product TypeGassing Manifold
DescriptionGassing manifold with 8 outlets
For Use With (Equipment)MaxQ 4000
Unit SizeEA