Benchtop Muffle Furnaces Accessories
Benchtop Muffle Furnaces Accessories
Actual product may vary
Benchtop Muffle Furnaces Accessories
Use these accessories with Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces.
Have Questions?
Catalog NumberDescriptionLength (English)
SH480X1
also known as AVL
Ceramic shelf6.87 in.
Catalog number SH480X1
also known as AVL
Price (MYR)
732.00
EA
Add to cart
Description:
Ceramic shelf
Length (English):
6.87 in.
Price (MYR)
732.00
EA
Add to cart
Use Thermo Scientific™ Benchtop Muffle Furnaces Accessories with Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces.
Specifications
DescriptionCeramic shelf
For Use With (Equipment)Thermolyne Benchtop Muffle Furnace
TypeCeramic Shelf
Dimensions (L x W x H) Exterior6.87 x 6.8 x 0.48 in. (17.4 x 17.3 x 1.2 cm)
Length (English)6.87 in.
Length (Metric)17.4 cm
Width (English)6.8 in.
Width (Metric)17.3 cm
Height (English)0.48 in.
Height (Metric)1.2 cm
Unit SizeEA