eVol™ Handheld Automated Analytical Syringe
eVol™ Handheld Automated Analytical Syringe
Actual product may vary
eVol™ Handheld Automated Analytical Syringe
eVol™ Handheld Automated Analytical Syringe
eVol™ Handheld Automated Analytical Syringe
Improves laboratory workflow
Have Questions?
Catalog NumberVolume (Metric)Needle Point StyleQuantity
SGE29100205 μLBevelEa.
Catalog number SGE2910020
Price (MYR)
792.00
PK
Add to cart
Volume (Metric):
5 μL
Needle Point Style:
Bevel
Quantity:
Ea.
Price (MYR)
792.00
PK
Add to cart
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen
Applications:
  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution
Specifications
Needle Gauge25 G
Diameter (English) Inner Needle0.0047 in.
Diameter (Metric) Inner Needle0.12 mm
Diameter (English) Outer Needle0.019 in.
Diameter (Metric) Outer Needle0.5 mm
For Use With (Application)Preparation of calibration standards, addition of internal standards
Length (English) Needle1.96 in.
Product TypeeVol Analytical Syringe
QuantityEa.
Volume (Metric)5 μL
Needle Point StyleBevel
For Use With (Equipment)eVol Automated Syringe
Unit SizePK