Human Integrin alpha 11 Antibody, R&D Systems™
Human Integrin alpha 11 Antibody, R&D Systems™
Actual product may vary
Human Integrin alpha 11 Antibody, R&D Systems™
Goat Polyclonal Antibody
Have Questions?
Catalog NumberQuantity
SGE18060002100 μg
Catalog number SGE18060002
Price (MYR)
796.00
EA
Add to cart
Quantity:
100 μg
Price (MYR)
796.00
EA
Add to cart
Integrin alpha 11 Polyclonal specifically detects Integrin alpha 11 in Human samples. It is validated for Western Blot.
Specifications
AntigenIntegrin alpha 11
ApplicationsWestern Blot
ClassificationPolyclonal
ConcentrationLYOPH
ConjugateUnconjugated
DilutionWestern Blot 1 ug/mL
Gene Accession No.NP_001004439
Gene AliasHsT18964, integrin alpha-11, integrin, alpha 11, ITGA11
Gene SymbolsITGA11
Host SpeciesGoat
ImmunogenChinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 11 Phe23-Pro1141 (Leu524Arg) Accession # NP_001004439
Purification MethodAffinity Purified
Quantity100 μg
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)22801
Test SpecificityDetects human Integrin alpha 11 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Integrin alpha 11 is observed and less than 1% cross-reactivity with recombinant human (rh) Integrin alpha 1 and rhIntegrin alpha 2 is observed.
ReconstitutionReconstitute at 0.2 mg/mL in sterile PBS.
Target SpeciesHuman
IsotypeIgG
Unit SizeEA
Contents & Storage
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.