Big Trap - universal
Big Trap - universal
Actual product may vary
Big Trap - universal
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
Have Questions?
Catalog NumberPort Type
SGE10353481/8 in.
Catalog number SGE1035348
Price (MYR)
2,875.00
EA
Add to cart
Port Type:
1/8 in.
Price (MYR)
2,875.00
EA
Add to cart
  • High capacity filters ideal for bulk purification - cumulative capacity level of 100μg/L of oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide at a flow rate of 8L/min
  • Efficiency < 100 ppb
  • Can be used where several instruments are plumbed from a single gas source
  • Sintered stainless steel frits prevent particulate contamination
For research use only
Specifications
For Use With (Application)GC and GCMS applications where helium is employed
ModelHydrocarbon
Volume (Metric)750 cm3
Port Type1/8 in.
DescriptionRemoves oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide
For Use WithBig Trap Mounting Clip
Pressure250 psig
Unit SizeEA