Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.
Have Questions?
Catalog number SGE1035220
Price (MYR)
1,439.00
EA
Add to cart
Price (MYR)
1,439.00
EA
Add to cart
Capacity of 7.2 g water for long filter lifetime
Indicator color change tells you when to change your filter
Can be used in combination with other filters through a simple connecting unit
For research use only
Specifications
Capacity (Metric)7.2 g H2O
Color IndicatorFrom green to pale brown
TypeMoisture
For Use With (Application)Removes water, oil and other foreign material from carrier gas