Check Order Status
Shimadzu Split / Splitless Liner with Middle Gooseneck
Actual product may vary
Shimadzu Split / Splitless Liner with Middle Gooseneck
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Have Questions?
Catalog Number
Product Type
Quantity
SGE092085
GC Inlet Liner
5/Pk.
Catalog number
SGE092085
Price (MYR)
971.00
PK
Add to cart
Product Type:
GC Inlet Liner
Quantity:
5/Pk.
Price (MYR)
971.00
PK
Add to cart
Each batch of liners tested for activity
Manufactured from borosilicate glass
Color coded by geometry for easy identification
For use with Shimazdu instruments
Specifications
Quantity
5/Pk.
Length (Metric)
95 mm
Product Type
GC Inlet Liner
Diameter (Metric) Inner
3.4 mm
Diameter (Metric) Outer
5 mm