Check Order Status
Shimadzu 14A Liners
Actual product may vary
Shimadzu 14A Liners
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Have Questions?
Catalog Number
Quantity
SGE0920831
5/Pk.
Catalog number
SGE0920831
Price (MYR)
971.00
PK
Add to cart
Quantity:
5/Pk.
Price (MYR)
971.00
PK
Add to cart
Each batch of liners tested for activity
Manufactured from borosilicate glass
Color coded by geometry for easy identification
For use with Shimazdu instruments
Specifications
Quantity
5/Pk.
Length (Metric)
99 mm
Product Type
GC Inlet liner, Split/Splitless Liner with Single Taper
Diameter (Metric) Inner
3.4 mm
Diameter (Metric) Outer
5 mm
For Use With (Equipment)
Shimadzu 14 A GC