Split / Splitless Tapered FocusLiner for Agilent
Split / Splitless Tapered FocusLiner for Agilent
Actual product may vary
Split / Splitless Tapered FocusLiner for Agilent
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Have Questions?
Catalog NumberQuantity
SGE09201125/Pk.
Catalog number SGE092011
Price (MYR)
5,171.00
PK
Add to cart
Quantity:
25/Pk.
Price (MYR)
5,171.00
PK
Add to cart
  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Agilent instruments
Specifications
Quantity25/Pk.
Length (Metric)78.5 mm
Product TypeGC Inlet Liner, Split/Splitless Tapered FocusLiner
Diameter (Metric) Inner4 mm
Diameter (Metric) Outer6.3 mm
For Use With (Equipment)Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
Unit SizePK