Check Order Status
SilTite™ Nuts for Mini Unions
SilTite™ Nuts for Mini Unions
SilTite Mini Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Have Questions?
Catalog Number
Packaging Quantity
SGE073553
5/Pk.
Catalog number
SGE073553
Price (MYR)
484.00
PK
Add to cart
Packaging Quantity:
5/Pk.
Price (MYR)
484.00
PK
Add to cart
Specifically designed for connecting capillary columns
Connects guard columns to capillary columns
Inert and precise glass lined bore
Low dead volume
Specifications
Type
Nut
Packaging Quantity
5/Pk.
Description
Nuts For Mini Unions of all ID