Replacement SilTite™ Mini Union Ferrules 
Replacement SilTite™ Mini Union Ferrules 
Actual product may vary
Replacement SilTite™ Mini Union Ferrules 
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Have Questions?
Catalog NumberFor Use With (Equipment)Quantity
SGE073470For 0.1 to 0.25 mm Columns10/Pk.
Catalog number SGE073470
Price (MYR)
526.00
PK
Add to cart
For Use With (Equipment):
For 0.1 to 0.25 mm Columns
Quantity:
10/Pk.
Price (MYR)
526.00
PK
Add to cart
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Diameter (Metric) Inner Tubing0.1 to 0.25 mm
For Use With (Equipment)For 0.1 to 0.25 mm Columns
TypeSilTite Mini Union Ferrules for 0.1 to 0.25 mm ID Columns
Quantity10/Pk.
Unit SizePK