15% Graphite/85% Vespel™ Reducing Ferrules
15% Graphite/85% Vespel™ Reducing Ferrules
Actual product may vary
15% Graphite/85% Vespel™ Reducing Ferrules
15% Graphite/85% Vespel™ Reducing Ferrules
15% Graphite/85% Vespel™ Reducing Ferrules
15% Graphite/85% Vespel™ Reducing Ferrules
Mechanically robust for long lifetimes
Have Questions?
Catalog NumberDiameter (Metric) InsideLength (Metric)Width (Metric)
SGE0726630.02 cm0.37 cm0.32 cm
Catalog number SGE072663
Price (MYR)
-
Diameter (Metric) Inside:
0.02 cm
Length (Metric):
0.37 cm
Width (Metric):
0.32 cm
Mechanically robust for long lifetimes
  • Form strong grip with capillary column
  • Maximum temperature: 300°C (572°F)
  • Must retighten after initial temperature cycles
  • Cannot be reused
  • Preconditioned under nitrogen for low-bleed performance
Specifications
Diameter (English) Inside0.0078 in.
For Use With (Application)GC
Length (Metric)0.37 cm
Width (English)0.12 in.
Width (Metric)0.32 cm
For Use With (Equipment)Agilent HP, Perkin Elmer, Varian GC/MS, Use with 0.0625 in. Swagelok, SGE Long Style SSNE Nuts
Material15% Graphite, 85% Vespel
TypeReducing Ferrule
Diameter (Metric) Inside0.02 cm
Unit SizePK