Check Order Status
SGE™ 100% Graphite Ferrules
Actual product may vary
SGE™ 100% Graphite Ferrules
Forms a stable seal with capillary columns
Have Questions?
Catalog Number
Diameter (Metric) Inside
SGE0726001
0.5 cm
Catalog number
SGE0726001
Price (MYR)
290.00
PK
Add to cart
Diameter (Metric) Inside:
0.5 cm
Price (MYR)
290.00
PK
Add to cart
Use in Agilent Technologies™ (HP) detectors and injectors at atmospheric pressure (e.g., FID)
Soft 100% graphite is porous to oxygen
Forms a stable seal with capillary columns
Reusable on other capillary columns
Maximum temperature: 450°C (842°F)
Preconditioned under nitrogen for low-bleed performance
SG-072635 and SG-072636 features short ferrule body style to fit standard HP injector/detector capillary nuts
Not for GC/MS interface
Specifications
Diameter (English) Inside
0.196 in.
Temperature (Metric) Max
450°C
For Use With (Equipment)
Injectors and Detectors at Atmospheric Pressure
Material
100% Graphite
Type
Ferrule
Diameter (Metric) Inside
0.5 cm