SolGel WAX Columns
SolGel WAX Columns
Actual product may vary
SolGel WAX Columns
Polyethylene Glycol (PEG) in a Sol-Gel matrix. Robust, high temperature polar WAX phase
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
SGE05478630 m0.53 mm0.5 μm
Catalog number SGE054786
Price (MYR)
4,448.00
EA
Add to cart
Length (Metric):
30 m
Diameter (Metric) Inner:
0.53 mm
Film Thickness:
0.5 μm
Price (MYR)
4,448.00
EA
Add to cart
  • Bonded polyethylene glycol
  • Very robust high-temperature column
  • Less susceptible to damage by oxygen than conventional wax phases
  • Polar phase
  • Low bleed and inert
Specifications
Functionality/Interaction TypeHydrogen Bonding or Dipole interactions
Diameter (Metric) Inner0.53 mm
Stationary PhasePolyethylene Glycol
USP TypeG14, G15, G16, G20, G39
Max. Temperature280°C
For Use With (Application)Acids Analysis, Amines Aliphatic, Amines Aromatic, Alcoholic Beverages Analysis, Essential Oils, Food - FAME, Phenols, Sugars-alditol Acetates
Temperature Range30°C to 260/280°C
Film Thickness0.5 μm
Length (Metric)30 m
Packing MaterialSolGel-Wax
PolarityPolar
CoatingFused Silica
Unit SizeEA