Check Order Status
BPX70 GC Capillary Column
Actual product may vary
BPX70 GC Capillary Column
SGE BPX70 GC Capillary Column is an MS premium, low bleed column with high polarity specifically designed for fatty acid methyl ester (FAME) analysis
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric) Inner
SGE054622
30 mm
0.25 mm
SGE054623
60 m
0.25 mm
2 Options
Catalog number
SGE054622
Price (MYR)
4,350.00
EA
Add to cart
Length (Metric):
30 mm
Diameter (Metric) Inner:
0.25 mm
Price (MYR)
4,350.00
EA
Add to cart
High temperature
Custom designed for separation of FAMEs
Ideal for cis/trans isomer separation
Polar phase
Specifications
Type
GC Capillary Column
Diameter (Metric) Inner
0.25 mm
Stationary Phase
BPX70
Max. Temperature
250/260°C
Temperature Range
50°C to 250/260°C
Film Thickness
0.25 mm
Length (Metric)
30 mm
Quantity
1 Each