Check Order Status
BP20 (WAX) Capillary Columns
Actual product may vary
BP20 (WAX) Capillary Columns
Industry standard WAX columns
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric) Inner
Film Thickness
SGE054438
30 m
0.32 mm
0.5 μm
SGE054427
30 m
0.25 mm
0.25 μm
SGE054433
30 m
0.32 mm
0.25 μm
3 Options
Catalog number
SGE054438
Price (MYR)
4,231.00
EA
Add to cart
Length (Metric):
30 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.5 μm
Price (MYR)
4,231.00
EA
Add to cart
Very polar polyethylene glycol (PEG) phase
Cross-linked for stability and washing
Also ideal for xylene and aromatic isomer
0.1 to 1μm film thickness: 20° to 260/280°C
Specifications
Diameter (Metric) Inner
0.32 mm
Stationary Phase
SGE™ BP20 (Wax)
For Use With (Application)
Ketones Analysis, Ketones, Aldehydes Analysis, Fragrance Analysis
Particle Size
0.5 μm
Film Thickness
0.5 μm
Length (Metric)
30 m
Polarity
Polar