BP5 Capillary GC Column
BP5 Capillary GC Column
Actual product may vary
BP5 Capillary GC Column
Excellent general purpose GC column
Have Questions?
Catalog NumberDiameter (Metric) Inner
SGE0541830.25 mm
Catalog number SGE054183
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Inner:
0.25 mm
  • Low bleed
  • Non-polar
  • Temperature Range: -60° to +320/340°C (-76°F to +608/644°F)
Specifications
Length (English)98 ft.
Diameter (Metric) Inner0.25 mm
Stationary PhaseSGE™ BP5
For Use With (Application)Hydrocarbon Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbon Analysis
Particle Size0.25 μm
Temperature Range-60°C to +320/340°C
Film Thickness0.25 μm
Length (Metric)30 m
Material5% Phenyl; 95% Dimethyl Polysiloxane
PolarityNon polar
Unit SizeEA