BPX5 GC Capillary Columns
BPX5 GC Capillary Columns
Actual product may vary
BPX5 GC Capillary Columns
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
SGE05411925 m0.32 mm0.25 μm
Catalog number SGE054119
Price (MYR)
4,433.00
EA
Add to cart
Length (Metric):
25 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.25 μm
Price (MYR)
4,433.00
EA
Add to cart
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Functionality/Interaction TypePi Bonding with the Aromatic Phenyl Groups
Diameter (Metric) Inner0.32 mm
Stationary Phase5% Phenyl Polysilphenylene Siloxane
USP TypeG27, G36, G41
Max. Temperature370°C
For Use With (Application)Phenols Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC/MS
Temperature Range-40°C to +360/370°C
Film Thickness0.25 μm
Length (Metric)25 m
Packing MaterialBPX5
PolarityNon polar
CoatingFused Silica
Unit SizeEA