BP1 GC Capillary Columns
BP1 GC Capillary Columns
Actual product may vary
BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
SGE05406230 m0.32 mm0.25 μm
Catalog number SGE054062
Price (MYR)
4,030.00
EA
Add to cart
Length (Metric):
30 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.25 μm
Price (MYR)
4,030.00
EA
Add to cart
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
TypeGC Column for Environmental Analysis
Diameter (Metric) Inner0.32 mm
Film Thickness0.25 μm
Length (Metric)30 m
Unit SizeEA