Replacement Needles for Luer-Lock Syringes
Replacement Needles for Luer-Lock Syringes
Actual product may vary
Replacement Needles for Luer-Lock Syringes
Replacement Needles for Luer-Lock Syringes
Replacement Needles for Luer-Lock Syringes
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Have Questions?
Catalog NumberNeedle GaugeLength (English) Needle
SGE03980723 G2.7 in.
Catalog number SGE039807
Price (MYR)
437.00
PK
Add to cart
Needle Gauge:
23 G
Length (English) Needle:
2.7 in.
Price (MYR)
437.00
PK
Add to cart
Designed for optimum septum penetration and prevention of septum coring.
  • Needle length is the exposed length of needle
  • Bevel tip style
  • Supplied in packs of 5
Specifications
MaterialStainless Steel
Needle Gauge23 G
Diameter (English) Inner0.012 in.
Diameter (English) Outer0.024 in.
Diameter (Metric) Inner0.32 mm
Diameter (Metric) Outer0.63 mm
For Use With (Application)Gas chromatography
Length (English) Needle2.7 in.
Product TypeReplacement Needle
Product LineTrajan
Length (Metric) Needle68.58 mm
Needle Point StyleBevel
Connection TypeLuer Lock
Unit SizePK