Syringe Plunger and Needle Kits
Syringe Plunger and Needle Kits
Actual product may vary
Syringe Plunger and Needle Kits
Syringe Plunger and Needle Kits
Syringe Plunger and Needle Kits
Designed to inject down to 1mL with high precision and accuracy
Have Questions?
Catalog NumberNeedle Gauge
SGE03911023 G
Catalog number SGE039110
Price (MYR)
335.00
PK
Add to cart
Needle Gauge:
23 G
Price (MYR)
335.00
PK
Add to cart
  • High precision and accuracy for small volume injections
  • Bevel Tip
  • For use with 1 to 2.5mL syringes
Specifications
Needle Gauge23 G
Diameter (English) Inner0.012 in.
Diameter (English) Outer0.024 in.
Diameter (Metric) Inner0.32 mm
Diameter (Metric) Outer0.63 mm
Length (English) Needle1.9 in.
Product TypeReplacement Syringe Plunger and Needle
Quantity5/Pk.
Needle Point StyleBevel
For Use With (Equipment)Syringe (1 to 2.5 mL)
Unit SizePK