Check Order Status
W-9 hydrochloride, Tocris Bioscience™
Element Search
Structure Search
W-9 hydrochloride, Tocris Bioscience™
Calmodulin antagonist
Have Questions?
Catalog Number
Quantity
SGE037010
10 mg
Catalog number
SGE037010
Price (MYR)
306.00
PK
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
306.00
PK
Add to cart
Specifications
Chemical Name or Material
W-9 hydrochloride
CAS
69762-85-2
Recommended Storage
Store at Room Temperature
Molecular Formula
C16H22Cl2N2O2S
Quantity
10 mg
View more
Calmodulin antagonist (inhibits calmodulin-activated PDE activity with an IC50 of 72 μM).