Check Order Status
Plunger Replacement for Thermo RSH and CTC RTC Autosampler Syringes
Plunger Replacement for Thermo RSH and CTC RTC Autosampler Syringes
SGE syringe replacement plungers for gas tight syringes
Have Questions?
Catalog Number
Volume (Metric)
SGE0355192
50 μL
Catalog number
SGE0355192
Price (MYR)
473.00
EA
Add to cart
Volume (Metric):
50 μL
Price (MYR)
473.00
EA
Add to cart
Replacement plungers for gas tight syringes
Designed to match specific syringe barrel to maintain gas tight seal
For sue with Thermo Sceintific instruments
Specifications
Material
PTFE (Plunger)
Product Type
RSH Autosampler Syringe
Volume (Metric)
50 μL
For Use With (Equipment)
Thermo RSH and CTC RTC Autosampler Syringe
Connection Type
Gas Tight