LC Pump Priming Syringe
LC Pump Priming Syringe
Actual product may vary
Thermo Scientific
LC Pump Priming Syringe
For loop fill injection or sample loop flush
Have Questions?
Catalog NumberVolume (Metric)
SGE0087625 mL
Catalog number SGE008762
Price (MYR)
479.00
EA
Add to cart
Volume (Metric):
5 mL
Price (MYR)
479.00
EA
Add to cart
  • Fixed Luer lock
Specifications
For Use With (Application)Used for loop fill injection or sample loop flush
Product TypeManual Syringe
Model5MDF-LL-GT
QuantityEa.
Volume (Metric)5 mL
Needle Point StyleLC
Unit SizeEA