Diamond Headspace Syringes for CTC PAL RTC (Robotic Tool Change)
Diamond Headspace Syringes for CTC PAL RTC (Robotic Tool Change)
Actual product may vary
Diamond Headspace Syringes for CTC PAL RTC (Robotic Tool Change)
Achieve outstanding reproducibility while saving time, and improving workflow.
Have Questions?
Catalog NumberVolume (Metric)Model
SGE0086552500 μL2.5MF-RTC/RSH-EGT-HS-6.5/0.63H
Catalog number SGE008655
Price (MYR)
1,564.00
EA
Add to cart
Volume (Metric):
2500 μL
Model:
2.5MF-RTC/RSH-EGT-HS-6.5/0.63H
Price (MYR)
1,564.00
EA
Add to cart
  • Reproducibility - a unique energized plunger tip design ensures a gas tight seal is consistently maintained providing outstanding performance and longevity with temperature cycling.
  • Operating temperature – premium performance continues even when temperature cycling up to 150°C.
  • Save time and improve workflow – extended equilibration times are not required. Reduced carryover minimizes the time used flushing and washing syringes.
Specifications
Needle Gauge23 G
Diameter (English) Outer Needle0.024 in.
Diameter (Metric) Outer Needle0.63 mm
For Use With (Application)Gas chromatography
Length (English) Needle2.5 in.
Product TypeDiamond Headspace Syringe
Model2.5MF-RTC/RSH-EGT-HS-6.5/0.63H
QuantityEa.
Volume (Metric)2500 μL
Needle Point StyleSide Hole
For Use With (Equipment)CTC PAL RTC
Connection TypeFixed
Unit SizeEA