SGE™ Syringes for Rheodyne and Valco Valves
SGE™ Syringes for Rheodyne and Valco Valves
Actual product may vary
SGE™ Syringes for Rheodyne and Valco Valves
Square cut needle with no electrotaper
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Connection Type
SGE007312500 μLRemovable
SGE005312100 μLRemovable
SGE005300100 μLFixed
Catalog number SGE007312
Price (MYR)
507.00
EA
Add to cart
Volume (Metric):
500 μL
Connection Type:
Removable
Price (MYR)
507.00
EA
Add to cart
Square cut needle with no electrotaper
  • Needles are 2 in., 22 gauge, 0.028 in. O.D.
Specifications
Accuracy±1%
Needle Gauge22 G
Diameter (English) Outer Needle0.028 in.
Diameter (Metric) Outer Needle0.71 mm
Length (English)2 in.
Length (English) Needle2 in.
Length (Metric)51 mm
Product TypeLC Manual Syringe with Needle
Max. Temperature (English)248°F
Max. Temperature (Metric)120°C
Model500R-GT-LC
QuantityEa.
Volume (Metric)500 μL
Product LineTrajan SGE
Length (Metric) Needle51 mm
Needle Point StyleBlunt
For Use With (Equipment)Rheodyne and Valco Valves
Connection TypeRemovable
Unit SizeEA