Gastight Syringes: Removable-Needle Models
Gastight Syringes: Removable-Needle Models
Actual product may vary
Gastight Syringes: Removable-Needle Models
Suitable for liquid and gas samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeDiameter (English) OuterNeedle Point Style
SGE005250100 μL25 G0.31 in.Removable Bevel
SGE0087005 mL23 G0.53 in.Removable Bevel
SGE00890010 mL23 G0.25 in.Removable Bevel
Catalog number SGE005250
Price (MYR)
491.00
EA
Add to cart
Volume (Metric):
100 μL
Needle Gauge:
25 G
Diameter (English) Outer:
0.31 in.
Needle Point Style:
Removable Bevel
Price (MYR)
491.00
EA
Add to cart
Suitable for liquid and gas samples
  • 10μL to 100mL capacities
  • Constructed from borosilicate glass, with PTFE plunger tips and Kel-F™ Luer cones (where applicable)
  • Needles align with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Replaceable plungers on all models
  • Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation; come with needle
Specifications
Accuracy±1%
MaterialBorosilicate Glass (Barrel), Borosilicate Glass (Barrel), PTFE (Tip), PTFE (Tip)
Needle Gauge25 G
Diameter (English) Outer Needle0.01 in.
Diameter (English) Outer0.31 in.
Diameter (Metric) Outer8 mm
Diameter (Metric) Outer Needle0.47 mm
For Use With (Application)Suitable for gases or liquids for maximum inertness
IncludesPlunger protection, needle glued into barrel with epoxy solvent-resistant glue
Length (English) Needle1.96 in.
Product TypeGastight Syringe
Model100R-GT
QuantityEa.
Volume (Metric)100 μL
Product LineSGE
Length (Metric) Needle50 mm
Needle Point StyleRemovable Bevel
Connection TypeRemovable