Series II Standard Fixed Needle Syringes
Series II Standard Fixed Needle Syringes
Actual product may vary
Series II Standard Fixed Needle Syringes
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeIncludesQuantity
SGE005000100 μL25 GStandard plungerEa.
SGE0010005 μL26 GPlunger protectionEa.
SGE00203010 μL26 GPlunger protection6 Pk.
SGE00400050 μL25 GStandard plungerEa.
SGE006000250 μL25 GStandard plungerEa.
SGE007000500 μL25 GStandard plungerEa.
Catalog number SGE005000
Price (MYR)
320.00
EA
Add to cart
Volume (Metric):
100 μL
Needle Gauge:
25 G
Includes:
Standard plunger
Quantity:
Ea.
Price (MYR)
320.00
EA
Add to cart
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Stainless-steel plungers are hand fitted, noninterchangeable, nonreplaceable
  • Syringes with 5 and 10μL capacities have barrel O.D. of 6.5mm, scale length of 54.1mm and include a 50mm long 26s gauge needle
  • 25 to 500μL capacity syringes have 8mm O.D. barrel, 60mm scale length and 50mm long, 25 gauge needle Fixed-needle models have needle glued into barrel with solvent-resistant epoxy glue Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation
Specifications
Accuracy±1%
MaterialBorosilicate Glass (Barrel), Borosilicate Glass (Barrel)
Needle Gauge25 G
Diameter (English) Outer Needle0.01 in.
Diameter (English) Outer0.31 in.
Diameter (Metric) Outer8 mm
Diameter (Metric) Outer Needle0.5 mm
IncludesStandard plunger
Length (English) Needle1.96 in.
Product TypeMicrovolume Syringe
GraduationsMajor - 10.0 μL, Minor - 2.0 μL
Model100F
QuantityEa.
Volume (Metric)100 μL
Product LineSGE
Length (Metric) Needle50 mm
Needle Point StyleFixed Bevel
Connection TypeFixed