Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Actual product may vary
Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Fixed needle replacement for manual syringes. For use with Beckman/Altex, Rheodyne, SSI, and Valco Valves
Have Questions?
Catalog NumberVolume (Metric)
SGE00430050 μL
Catalog number SGE004300
Price (MYR)
308.00
EA
Add to cart
Volume (Metric):
50 μL
Price (MYR)
308.00
EA
Add to cart
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
MaterialBorosilicate Glass (Barrel)
Needle Gauge22 G
Diameter (Metric) Inner Needle0.37 mm
Diameter (Metric) Outer0.71 mm
Diameter (Metric) Outer Needle0.028 mm
Product TypeManual Syringe
Volume (Metric)50 μL
Length (Metric) Needle51 mm
Needle Point StyleLC
For Use With (Equipment)Beckman/Altex, Rheodyne, SSI, Valco Valves
Unit SizeEA