PTFE Stir Bar Retriever
PTFE Stir Bar Retriever
Actual product may vary
PTFE Stir Bar Retriever
Use to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.
Have Questions?
Catalog NumberLength (Metric)
SGE004250250 mm
Catalog number SGE004250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Use to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.
  • Inert and leak-free
  • Molded in pure, FDA-approved PTFE
  • Alnico V magnets
  • Autoclavable
Specifications
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Application)Removal of Stirrer Bars From Vessels
For Use With (Equipment)PTFE Stirrer Bars
Height (Metric)10 mm
CoatingPTFE
Diameter (Metric)10 mm
Length (Metric)250 mm
MaterialPTFE with Alnico V Magnet Core
Quantity1/Ea.
ShapeRetriever
TypeStir Bar Retriever
Unit SizeEA