PTFE Stirring Rod with Steel Core
PTFE Stirring Rod with Steel Core
Actual product may vary
PTFE Stirring Rod with Steel Core
Can be used in temperatures up to 280°C (540°F). The PTFE stirring rod has a steel core. The rod can be bent to a permanent shape.
Have Questions?
Catalog NumberLength (Metric)
SGE003250250 mm
Catalog number SGE003250
Price (MYR)
450.00
EA
Add to cart
Length (Metric):
250 mm
Price (MYR)
450.00
EA
Add to cart
  • Chemically resistant, inert, and leak-free
  • Can be used in temperatures up to 280°C (540°F)
  • Steel core for maximum rigidity
  • Can be bent in to a permanent shape
  • Will not scratch
Specifications
AutoclavableAutoclavable
DescriptionPTFE encapsulated Mild Steel Core
For Use With (Application)Mixing of various chemicals
Weight (Metric)35g
Diameter (Metric)6 mm
Length (Metric)250 mm
Quantity1/Ea.
Unit SizeEA