Thermo/PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Thermo/PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Actual product may vary
Thermo/PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Features Diamond Syringe Technology which results in improved solvent resistance, wide temperature ranges, and increased operational smoothness. SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle are used for CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus, AS2000, AS3000, AS200/800 Autosamplers.
Have Questions?
Catalog NumberVolume (Metric)Needle GaugeDiameter (Metric) Outer NeedleModel
SGE00298010 μL26 G0.47 mm10F-C/T-5/0.47C
Catalog number SGE002980
Price (MYR)
361.00
EA
Add to cart
Volume (Metric):
10 μL
Needle Gauge:
26 G
Diameter (Metric) Outer Needle:
0.47 mm
Model:
10F-C/T-5/0.47C
Price (MYR)
361.00
EA
Add to cart
  • Improved solvent resistance
  • Wide temperature ranges up to 150°C
  • Increased operational smoothness
  • Designed specifically for the PAL
  • 22 gauge needles are 0.72mm OD
  • 23 gauge needles are 0.63mm OD
  • 26 gauge needles are 0.47mm OD
  • Accuracy and Reproducibility;  ±1% (dispensed volume) (±2% for 0.5μL syringes)
  • Cone shaped (also known as point style AS for autosampler), are designed to withstand multi injection demands and improves septum lifetime when used with an autosampler
  • Gas tight plunger, where applicable, is made of PTFE
Specifications
Needle Gauge26 G
Diameter (Metric) Outer Needle0.47 mm
For Use With (Application)Gas chromatography
Length (Metric)50 mm
Product TypeAutosampler Syringe
Model10F-C/T-5/0.47C
Volume (Metric)10 μL
Needle Point StyleCone
For Use With (Equipment)CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus and AS3000, AS2000, AS200/800
Connection TypeFixed
Unit SizeEA